missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyridine, 98+%

CAS: 81565-18-6 | C6H3ClF3N | 181.54 g/mol

Supplier:  Thermo Scientific Chemicals L1071303

 View more versions of this product

Catalog No. AAL1071303


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-4-(trifluoromethyl)pyridine
1.411
40°C (104°F)
1.449
1 g
4246276
GBNPVXZNWBWNEN-UHFFFAOYSA-N
2-chloro-4-(trifluoromethyl)pyridine
144958
≥98%
81565-18-6
145°C
C6H3ClF3N
MFCD00042224
UN1992
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
FC(F)(F)C1=CC=NC(Cl)=C1
181.54
181.55

Chemical Identifiers

81565-18-6
181.54
GBNPVXZNWBWNEN-UHFFFAOYSA-N
144958
FC(F)(F)C1=CC=NC(Cl)=C1
C6H3ClF3N
MFCD00042224
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
2-chloro-4-(trifluoromethyl)pyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only