missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. C17905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17905G 5 g
C17901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17905G Supplier TCI America Supplier No. C17905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175205-54-6
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.05
MDL Number MFCD00052912
InChI Key NJXDBSSSDPOAFI-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
PubChem CID 2736675
IUPAC Name 2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
SMILES FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 279.05
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzenesulfonyl Chloride