missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)aniline, 98%

Catalog No. p-7082013
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1224206 5 g
AAA1224214 25 g
AAA1224222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1224206 Supplier Thermo Scientific Chemicals Supplier No. A1224206

CAS: 39885-50-2 | C7H5ClF3N | 195.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39885-50-2
Molecular Formula C7H5ClF3N
Molecular Weight (g/mol) 195.57
MDL Number MFCD00042563
InChI Key MBBUTABXEITVNY-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl aniline, 4-amino-3-chlorobenzotrifluoride, 4-amino-5-chloro-benzotrifluoride, benzenamine, 2-chloro-4-trifluoromethyl, 3-chloro-4-aminobenzotrifluoride, 2-chloro-4-trifluoromethyl-phenylamine, 2-chloro-4-trifluoromethyl benzenamine, 2-chloro-4-trifluoromethyl phenylamine, 4-amino-3-chloro-benzotrifluoride, pubchem2474
PubChem CID 162001
IUPAC Name 2-chloro-4-(trifluoromethyl)aniline
SMILES NC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Density 1.434
Boiling Point 214°C to 218°C
Flash Point 103°C (217°F)
Refractive Index 1.5015
Quantity 5 g
UN Number UN2810
Beilstein 4179703
Formula Weight 195.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4- (trifluoromethyl)aniline
Hazard Category H301+H311-H315-H319-H332-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 254-674-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only