missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(trifluoromethoxy)aniline 97.0+%, TCI America™
$83.89 - $247.92
Chemical Identifiers
| CAS | 69695-61-0 |
|---|---|
| Molecular Formula | C7H5ClF3NO |
| Molecular Weight (g/mol) | 211.568 |
| MDL Number | MFCD01631704 |
| InChI Key | YMLWONHDNGICOH-UHFFFAOYSA-N |
| PubChem CID | 2734084 |
| IUPAC Name | 2-chloro-4-(trifluoromethoxy)aniline |
| SMILES | C1=CC(=C(C=C1OC(F)(F)F)Cl)N |
Chemical Identifiers
| 69695-61-0 | |
| 211.568 | |
| YMLWONHDNGICOH-UHFFFAOYSA-N | |
| 2-chloro-4-(trifluoromethoxy)aniline |
| C7H5ClF3NO | |
| MFCD01631704 | |
| 2734084 | |
| C1=CC(=C(C=C1OC(F)(F)F)Cl)N |
Specifications
| 69695-61-0 | |
| 96°C | |
| Clear Liquid at 20°C | |
| MFCD01631704 | |
| YMLWONHDNGICOH-UHFFFAOYSA-N | |
| 2-chloro-4-(trifluoromethoxy)aniline | |
| 2734084 | |
| ≥97.0% (GC,T) |
| Yellow | |
| 1 g | |
| C7H5ClF3NO | |
| 2-chloro-4-trifluoromethoxy aniline, benzenamine, 2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy phenylamine, benzenamine,2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy-phenylamine, pubchem2941, acmc-209wey, ksc630s5l, 2-chloro4-trifluoromethoxyaniline, 4-trifluoromethoxy-2-chloroaniline | |
| C1=CC(=C(C=C1OC(F)(F)F)Cl)N | |
| 211.568 | |
| 211.57 | |
| 2-Chloro-4-(trifluoromethoxy)aniline |
Safety and Handling
TSCA : No