missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(trifluoromethoxy)aniline 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Physical Form | Clear Liquid at 20°C |
| Synonym | 2-chloro-4-trifluoromethoxy aniline, benzenamine, 2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy phenylamine, benzenamine,2-chloro-4-trifluoromethoxy, 2-chloro-4-trifluoromethoxy-phenylamine, pubchem2941, acmc-209wey, ksc630s5l, 2-chloro4-trifluoromethoxyaniline, 4-trifluoromethoxy-2-chloroaniline |
| Boiling Point | 96°C |
| Formula Weight | 211.57 |
| Percent Purity | ≥97.0% (GC,T) |
| Chemical Name or Material | 2-Chloro-4-(trifluoromethoxy)aniline |
Safety and Handling
| TSCA | No |