missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloro-4-pyridinyl)methanol, 97%, Thermo Scientific™

Catalog No. p-3522358
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53609DA 1 g Amber Glass Bottle
CC53609DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53609DA Supplier Thermo Scientific Chemicals Supplier No. CC53609DA
Only null left

Chemical Identifiers

CAS 100704-10-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD06858767
InChI Key UDDVPFLXGOBESH-UHFFFAOYSA-N
Synonym 2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol
PubChem CID 7062237
IUPAC Name (2-chloropyridin-4-yl)methanol
SMILES C1=CN=C(C=C1CO)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 143.57
Percent Purity 95%
Chemical Name or Material (2-Chloro-4-pyridinyl)methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.