accessibility menu, dialog, popup

UserName

HONCl

CAS RN 100704-10-7

IUPAC Name: (2-chloropyridin-4-yl)methanol
Synonyms: (2-chloropyridin-4-yl)methanol
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: UDDVPFLXGOBESH-UHFFFAOYSA-N
SMILES: OCC1=CC=NC(Cl)=C1
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)