missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. C27231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27231G 1 g
C27235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27231G Supplier TCI America Supplier No. C27231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101066-61-9
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.55
MDL Number MFCD06651557
InChI Key UFPOSTQMFOYHJI-UHFFFAOYSA-N
Synonym 2-chloroisonicotinaldehyde, 2-chloro-4-pyridinecarboxaldehyde, 2-chloro-4-formylpyridine, 2-chloro-pyridine-4-carbaldehyde, 2-chloropyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde, 2-chloro, pubchem5099, 6-chloroisonicotinaldehyde, ksc180g9n, 2-chloropyridin-4-carbaldehyde
PubChem CID 2762994
IUPAC Name 2-chloropyridine-4-carbaldehyde
SMILES ClC1=CC(C=O)=CC=N1

Specifications

Melting Point 49°C
Color White
Quantity 1 g
Formula Weight 141.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-pyridinecarboxaldehyde