missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-phenylquinazoline 98.0+%, TCI America™

Catalog No. p-7125871
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31731G 1 g
C31735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31731G Supplier TCI America Supplier No. C31731G

Chemical Identifiers

CAS 29874-83-7
Molecular Formula C14H9ClN2
Molecular Weight (g/mol) 240.69
MDL Number MFCD01152724
InChI Key SFKMVPQJJGJCMI-UHFFFAOYSA-N
PubChem CID 3123582
IUPAC Name 2-chloro-4-phenylquinazoline
SMILES ClC1=NC(C2=CC=CC=C2)=C2C=CC=CC2=N1

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
Formula Weight 240.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-phenylquinazoline