accessibility menu, dialog, popup

UserName

CAS RN 29874-83-7

ClNN

CAS RN 29874-83-7

IUPAC Name: 2-chloro-4-phenylquinazoline
Synonyms: 2-chloro-4-phenylquinazoline
Molecular Weight (g/mol): 240.69
Molecular Formula: C14H9ClN2
InChi Key: SFKMVPQJJGJCMI-UHFFFAOYSA-N
SMILES: ClC1=NC(C2=CC=CC=C2)=C2C=CC=CC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results