missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrotoluene 99.0+%, TCI America™

Catalog No. C069025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C069025G 25 g
C0690500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C069025G Supplier TCI America Supplier No. C069025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-86-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007210
InChI Key LLYXJBROWQDVMI-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrotoluene, benzene, 2-chloro-1-methyl-4-nitro, o-chloro-p-nitrotoluol, toluene, 2-chloro-4-nitro, 3-chloro-4-methylnitrobenzene, 2-chlor-4-nitrotoluen, unii-9t26lnr6t8, 2-chloro-4-nitrotoluen czech, 2,4-chloronitrotoluene, 2-chloro-4-nitro-1-methylbenzene
PubChem CID 8491
IUPAC Name 2-chloro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 63°C
Color Yellow
Boiling Point 260°C
Quantity 25 g
UN Number 3457
Formula Weight 171.58
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitrotoluene
EINECSNumber (3)-0449
RTECSNumber XS9100000
TSCA Yes