missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrotoluene, 98%

Catalog No. AAB2354214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2354214 25 g
AAB2354222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2354214 Supplier Thermo Scientific Chemicals Supplier No. B2354214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 121-86-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-86-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007210
InChI Key LLYXJBROWQDVMI-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrotoluene, benzene, 2-chloro-1-methyl-4-nitro, o-chloro-p-nitrotoluol, toluene, 2-chloro-4-nitro, 3-chloro-4-methylnitrobenzene, 2-chlor-4-nitrotoluen, unii-9t26lnr6t8, 2-chloro-4-nitrotoluen czech, 2,4-chloronitrotoluene, 2-chloro-4-nitro-1-methylbenzene
PubChem CID 8491
IUPAC Name 2-chloro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 62°C to 66°C
Boiling Point 260°C
Quantity 25 g
UN Number UN3457
Beilstein 1817924
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID
EINECSNumber 204-501-7
RTECSNumber XS9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only