missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrophenyl isothiocyanate, 97%

Catalog No. AAL1089803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1089803 1 g
AAL1089806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1089803 Supplier Thermo Scientific Chemicals Supplier No. L1089803
Only null left

CAS: 23165-64-2 | C7H3ClN2O2S | 214.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23165-64-2
Molecular Formula C7H3ClN2O2S
Molecular Weight (g/mol) 214.62
MDL Number MFCD00041244
InChI Key JSYNALWWKRNZDX-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrophenyl isothiocyanate, 2-chloro-4-nitrophenylisothiocyanate, benzene,2-chloro-1-isothiocyanato-4-nitro, 2-chloro-4-nitrobenzenisothiocyanate, acmc-1cg9c, 2-chloranyl-1-isothiocyanato-4-nitro-benzene
PubChem CID 2757736
IUPAC Name 2-chloro-1-isothiocyanato-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=S

Specifications

Melting Point 95°C to 99°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2972987
Sensitivity Moisture sensitive
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-nitrophenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only