missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrophenol 98.0+%, TCI America™

Catalog No. C022725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022725G 25 g
C0227500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C022725G Supplier TCI America Supplier No. C022725G

Chemical Identifiers

CAS 619-08-9
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD00043910
InChI Key BOFRXDMCQRTGII-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
PubChem CID 12074
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)O

Specifications

Melting Point 110°C
Quantity 25 g
Formula Weight 173.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitrophenol
RTECSNumber SK5075000
TSCA Yes