missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrophenol, 97%

CAS: 619-08-9 | C6H4ClNO3 | 173.552 g/mol

Supplier:  Thermo Scientific Chemicals B2156118

 View more versions of this product

Catalog No. AAB2156118


Only null left
Explore available promotions

Description

Description

2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.

Solubility
Slightly Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Specifications

Specifications

2-Chloro-4-nitrophenol
108°C to 112°C
C6H4ClNO3
50 g
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
BOFRXDMCQRTGII-UHFFFAOYSA-N
173.552
173.56
619-08-9
Characteristic
MFCD00043910
2046372
Slightly Soluble in water.
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O
12074
97%

Chemical Identifiers

619-08-9
173.552
BOFRXDMCQRTGII-UHFFFAOYSA-N
12074
C6H4ClNO3
MFCD00043910
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-578-8

RTECSNumber : SK5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only