missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzoyl Chloride 98.0+%, TCI America™

Catalog No. C042525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C042525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C042525G Supplier TCI America Supplier No. C042525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7073-36-1
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.01
MDL Number MFCD00051515
InChI Key KTHNITVDTYAHFF-UHFFFAOYSA-N
Synonym benzoyl chloride, 2-chloro-4-nitro, benzoyl chloride,2-chloro-4-nitro, acmc-209ofu, 2-chloro-4-nitro-benzoyl chloride
PubChem CID 81522
IUPAC Name 2-chloro-4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
UN Number 3261
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-4-nitrobenzoyl Chloride