missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzoyl chloride, 98%

Catalog No. AAL0824606
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0824606 5 g
AAL0824618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0824606 Supplier Thermo Scientific Chemicals Supplier No. L0824606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7073-36-1 | C7H3Cl2NO3 | 220.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7073-36-1
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.01
MDL Number MFCD00051515
InChI Key KTHNITVDTYAHFF-UHFFFAOYSA-N
PubChem CID 81522
IUPAC Name 2-chloro-4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1

Specifications

Boiling Point 165°C (20 mmHg)
Refractive Index 1.595
Quantity 5 g
UN Number UN3265
Beilstein 519794
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 2-chloro-4-nitro, benzoyl chloride,2-chloro-4-nitro, acmc-209ofu, 2-chloro-4-nitro-benzoyl chloride
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 230-367-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only