missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-nitrobenzoic Acid 98.0+%, TCI America™
$42.87 - $221.83
Chemical Identifiers
| CAS | 99-60-5 |
|---|---|
| Molecular Formula | C7H4ClNO4 |
| Molecular Weight (g/mol) | 201.562 |
| MDL Number | MFCD00007209 |
| InChI Key | QAYNSPOKTRVZRC-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde |
| PubChem CID | 7448 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O |
Chemical Identifiers
| 99-60-5 | |
| 201.562 | |
| QAYNSPOKTRVZRC-UHFFFAOYSA-N | |
| 7448 |
| C7H4ClNO4 | |
| MFCD00007209 | |
| 2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O |
Specifications
| 99-60-5 | |
| White-Yellow | |
| MFCD00007209 | |
| 2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde | |
| C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O | |
| 7448 | |
| ≥98.0% (GC,T) | |
| 2-Chloro-4-nitrobenzoic Acid |
| 138°C | |
| C7H4ClNO4 | |
| 25 g | |
| QAYNSPOKTRVZRC-UHFFFAOYSA-N | |
| 201.562 | |
| 201.56 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1502
RTECSNumber : DG5425000
TSCA : Yes