missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C076425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C076425G 25 g
C0764500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C076425G Supplier TCI America Supplier No. C076425G
Only null left

Chemical Identifiers

CAS 99-60-5
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007209
InChI Key QAYNSPOKTRVZRC-UHFFFAOYSA-N
Synonym 2-chloro-4-nitrobenzoic acid, benzoic acid, 2-chloro-4-nitro, kyselina 2-chloro-4-nitrobenzoova, kyselina 2-chloro-4-nitrobenzoova czech, 2-chloro-4-nitrobenzoic, 2-chloro-4-nitro-benzoic acid, 2-chloro-4-nitro benzoic acid, 2-chloro-4-nitrobenzoicacid, pubchem20257, acmc-209sde
PubChem CID 7448
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)O

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitrobenzoic Acid
EINECSNumber (3)-1502
RTECSNumber DG5425000
TSCA Yes