missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylsulphonylbenzoic acid, 95%, Thermo Scientific™

$61.88 - $61.88

Chemical Identifiers

CAS 53250-83-2
Molecular Formula C8H6ClO4S
Molecular Weight (g/mol) 233.64
MDL Number MFCD00216496
InChI Key CTTWSFIIFMWHLQ-UHFFFAOYSA-M
Synonym 2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
PubChem CID 735863
IUPAC Name 2-chloro-4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC425010010
SDS View Documents
Thermo Scientific Chemicals
425010010
1 g Glass bottle
Each for $61.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

53250-83-2
233.64
CTTWSFIIFMWHLQ-UHFFFAOYSA-M
735863
CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1
C8H6ClO4S
MFCD00216496
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
2-chloro-4-methylsulfonylbenzoic acid

Specifications

53250-83-2
100.0
Pink or White to Yellow
Authentic
Glass bottle
MFCD00216496
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
CTTWSFIIFMWHLQ-UHFFFAOYSA-M
2-chloro-4-methylsulfonylbenzoic acid
735863
95%
2-Chloro-4-methylsulphonylbenzoic acid, 95%
94.0
192.0°C to 193.0°C
5% max.
95%
C8H6ClO4S
1 g
Solubility in water: insoluble
CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1
233.64
234.66
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only