missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxyphenol 98.0+%, TCI America™

Catalog No. p-7125010
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C14875G 5 g
C148725G 25 g
2 options

Catalog No. C14875G

Supplier: TCI America C14875G

Only null left

Chemical Identifiers

CAS 18113-03-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00070773
InChI Key GNVRRKLFFYSLGT-UHFFFAOYSA-N
PubChem CID 87459
IUPAC Name 2-chloro-4-methoxyphenol
SMILES COC1=CC(=C(C=C1)O)Cl

Specifications

Melting Point 43°C
Quantity 5 g
Synonym 3-chloro-4-hydroxyanisole, phenol, chloro-4-methoxy, phenol, 2-chloro-4-methoxy, pubchem3663, 2-chloro-4-methoxy-phenol, acmc-1c55n, phenol,2-chloro-4-methoxy, ksc445k0p, 2-chloro-4-methoxyphenol
Formula Weight 158.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methoxyphenol
TSCA No
Recommended Storage Refrigerator