missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$108.11 - $108.11
Specifications
| CAS | 603122-80-1 |
|---|---|
| Color | White-Yellow |
| Molecular Formula | C8H8BClO4 |
| MDL Number | MFCD08689530 |
| Synonym | 2-Chloro-4-(methoxycarbonyl)benzeneboronic Acid |
Specifications
| 603122-80-1 | |
| C8H8BClO4 | |
| 2-Chloro-4-(methoxycarbonyl)benzeneboronic Acid | |
| 22171291 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD08689530 | |
| ITEQHBSWRMYSRP-UHFFFAOYSA-N | |
| 214.41 |