missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C32241G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32241G Supplier TCI America Supplier No. C32241G

Specifications

Chemical Name or Material 2-Chloro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 603122-80-1
Color White-Yellow
Molecular Formula C8H8BClO4
Quantity 1 g
MDL Number MFCD08689530
Synonym 2-Chloro-4-(methoxycarbonyl)benzeneboronic Acid
InChI Key ITEQHBSWRMYSRP-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(Cl)=C(C=C1)B(O)O
IUPAC Name [2-chloro-4-(methoxycarbonyl)phenyl]boronic acid
Molecular Weight (g/mol) 214.41
PubChem CID 22171291
Formula Weight 214.41
Physical Form Crystalline Powder
Show More Show Less