missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-iodobenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 167°C |
| Quantity | 5 g |
| Formula Weight | 282.46 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-4-iodobenzoic Acid |
Safety and Handling
| TSCA | No |