missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodoaniline 98.0+%, TCI America™

Catalog No. C17815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17815G Supplier TCI America Supplier No. C17815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42016-93-3
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00025298
InChI Key MYDAOWXYGPEPJT-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-phenylamine, benzenamine, 2-chloro-4-iodo, 2-chloro-4-iodo-aniline, ccris 9438, zlchem 542, pubchem3811, 2-chloro-4-iodoanilline, 2-chloro-4-iodo aniline, acmc-1am5p, 2-chloro-4-iodophenylamine
PubChem CID 282930
IUPAC Name 2-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C=C1Cl

Specifications

Melting Point 72°C
Quantity 5 g
Formula Weight 253.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-iodoaniline