missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodoaniline, 98%

Catalog No. AAA1074206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1074206 5 g
AAA1074214 25 g
AAA1074222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1074206 Supplier Thermo Scientific Chemicals Supplier No. A1074206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 42016-93-3 | C6H5ClIN | 253.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42016-93-3
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00025298
InChI Key MYDAOWXYGPEPJT-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-phenylamine, benzenamine, 2-chloro-4-iodo, 2-chloro-4-iodo-aniline, ccris 9438, zlchem 542, pubchem3811, 2-chloro-4-iodoanilline, 2-chloro-4-iodo aniline, acmc-1am5p, 2-chloro-4-iodophenylamine
PubChem CID 282930
IUPAC Name 2-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C=C1Cl

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
UN Number UN2811
Beilstein 2081106
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-iodoaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only