missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-iodo-3-methylpyridine 98.0+%, TCI America™
Supplier: TCI America C33685G
Specifications
| 2-Chloro-4-iodo-3-methylpyridine | |
| 106°C | |
| MFCD03095293 | |
| 2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline | |
| CC1=C(C=CN=C1Cl)I | |
| 253.467 | |
| 253.47 | |
| Crystalline Powder |
| 153034-88-9 | |
| C6H5ClIN | |
| 5 g | |
| WHCRABJNJRAJIP-UHFFFAOYSA-N | |
| 2-chloro-4-iodo-3-methylpyridine | |
| 10083628 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 153034-88-9 | |
| 253.467 | |
| WHCRABJNJRAJIP-UHFFFAOYSA-N | |
| 10083628 | |
| CC1=C(C=CN=C1Cl)I |
| C6H5ClIN | |
| MFCD03095293 | |
| 2-chloro-4-iodo-3-picoline, 2-chloro-3-methyl-4-iodopyridine, 2-chloro-4-iodo-3-methyl-pyridine, pyridine, 2-chloro-4-iodo-3-methyl, 2-chloro-4-iodo-3-methylpyridne, pubchem17059, 2-chloranyl-4-iodanyl-3-methyl-pyridine, 2-chloro-4-iodo-3-methylpyridine, 2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline | |
| 2-chloro-4-iodo-3-methylpyridine |
Safety and Handling
TSCA : No