accessibility menu, dialog, popup

UserName

CAS RN 153034-88-9

CH3NClI

CAS RN 153034-88-9

IUPAC Name: 2-chloro-4-iodo-3-methylpyridine
Synonyms: 2-chloro-4-iodo-3-methylpyridine
Molecular Weight (g/mol): 253.47
Molecular Formula: C6H5ClIN
InChi Key: WHCRABJNJRAJIP-UHFFFAOYSA-N
SMILES: CC1=C(I)C=CN=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)