missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorophenylacetic acid, 97%

Catalog No. AAB2287214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2287214 25 g
AAB2287206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2287214 Supplier Thermo Scientific Chemicals Supplier No. B2287214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 177985-32-9 | C8H6ClFO2 | 188.582 g/mol

2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 177985-32-9
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00236028
InChI Key CDUSPKFHAVQVRQ-UHFFFAOYSA-N
PubChem CID 2725061
IUPAC Name 2-(2-chloro-4-fluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1F)Cl)CC(=O)O

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
Synonym 2-chloro-4-fluorophenylacetic acid, 2-2-chloro-4-fluorophenyl acetic acid, 2-chloro-4-fluorophenyl acetic acid, 2-2-chloro-4-fluoro-phenyl acetic acid, rarechem al bo 0843, timtec-bb sbb003675, labotest-bb lt00441150, benzeneacetic acid,2-chloro-4-fluoro, 2-chloro-4-fluoro-phenyl-acetic acid, benzeneacetic acid, 2-chloro-4-fluoro
Solubility Information Slightly soluble in water.
Formula Weight 188.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only