missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluorobenzyl Bromide 98.0+%, TCI America™
$77.00 - $225.25
Chemical Identifiers
| CAS | 45767-66-6 |
|---|---|
| Molecular Formula | C7H5BrClF |
| Molecular Weight (g/mol) | 223.469 |
| MDL Number | MFCD00236025 |
| InChI Key | GAUUDQVOPUKGJD-UHFFFAOYSA-N |
| Synonym | 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide |
| PubChem CID | 2725062 |
| IUPAC Name | 1-(bromomethyl)-2-chloro-4-fluorobenzene |
| SMILES | C1=CC(=C(C=C1F)Cl)CBr |
Chemical Identifiers
| 45767-66-6 | |
| 223.469 | |
| GAUUDQVOPUKGJD-UHFFFAOYSA-N | |
| 2725062 | |
| C1=CC(=C(C=C1F)Cl)CBr |
| C7H5BrClF | |
| MFCD00236025 | |
| 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide | |
| 1-(bromomethyl)-2-chloro-4-fluorobenzene |
Specifications
| 45767-66-6 | |
| C7H5BrClF | |
| 5 g | |
| 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide | |
| C1=CC(=C(C=C1F)Cl)CBr | |
| 223.469 | |
| 223.47 | |
| Crystalline Powder |
| 39°C | |
| MFCD00236025 | |
| 3261 | |
| GAUUDQVOPUKGJD-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2-chloro-4-fluorobenzene | |
| 2725062 | |
| ≥98.0% (GC) | |
| 2-Chloro-4-fluorobenzyl Bromide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator