missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluorobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America C269725G
Specifications
| 2-Chloro-4-fluorobenzoyl Chloride | |
| Yellow | |
| C7H3Cl2FO | |
| 25 g | |
| 2-chloro-4-fluorobenzoylchloride, 2-chloro-4-fluoro-benzoyl chloride, 2-chloro-4-fluorobenzene-1-carbonyl chloride, benzoyl chloride, 2-chloro-4-fluoro, benzoyl chloride,2-chloro-4-fluoro, acmc-20anzq, ksc547s3d, 2-chlor-4-fluorbenzoylchlorid, 2chloro-4-fluorobenzoyl chloride, 4-fluoro-2-chlorobenzoyl chloride | |
| C1=CC(=C(C=C1F)Cl)C(=O)Cl | |
| 192.998 | |
| 193.00 | |
| Liquid |
| 21900-54-9 | |
| 95°C | |
| MFCD00084954 | |
| 3265 | |
| POIAZJJVWRVLBO-UHFFFAOYSA-N | |
| 2-chloro-4-fluorobenzoyl chloride | |
| 2736546 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 21900-54-9 | |
| 192.998 | |
| POIAZJJVWRVLBO-UHFFFAOYSA-N | |
| 2736546 | |
| C1=CC(=C(C=C1F)Cl)C(=O)Cl |
| C7H3Cl2FO | |
| MFCD00084954 | |
| 2-chloro-4-fluorobenzoylchloride, 2-chloro-4-fluoro-benzoyl chloride, 2-chloro-4-fluorobenzene-1-carbonyl chloride, benzoyl chloride, 2-chloro-4-fluoro, benzoyl chloride,2-chloro-4-fluoro, acmc-20anzq, ksc547s3d, 2-chlor-4-fluorbenzoylchlorid, 2chloro-4-fluorobenzoyl chloride, 4-fluoro-2-chlorobenzoyl chloride | |
| 2-chloro-4-fluorobenzoyl chloride |
Safety and Handling
TSCA : No