missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. C26975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26975G 5 g
C269725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26975G Supplier TCI America Supplier No. C26975G

Chemical Identifiers

CAS 21900-54-9
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00084954
InChI Key POIAZJJVWRVLBO-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzoylchloride, 2-chloro-4-fluoro-benzoyl chloride, 2-chloro-4-fluorobenzene-1-carbonyl chloride, benzoyl chloride, 2-chloro-4-fluoro, benzoyl chloride,2-chloro-4-fluoro, acmc-20anzq, ksc547s3d, 2-chlor-4-fluorbenzoylchlorid, 2chloro-4-fluorobenzoyl chloride, 4-fluoro-2-chlorobenzoyl chloride
PubChem CID 2736546
IUPAC Name 2-chloro-4-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1F)Cl)C(=O)Cl

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 3265
Formula Weight 193.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-fluorobenzoyl Chloride