missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4'-fluorobenzophenone 98.0+%, TCI America™
$137.94 - $411.95
Chemical Identifiers
| CAS | 1806-23-1 |
|---|---|
| Molecular Formula | C13H8ClFO |
| Molecular Weight (g/mol) | 234.654 |
| MDL Number | MFCD00000559 |
| InChI Key | DODIKYQYCCFWRZ-UHFFFAOYSA-N |
| PubChem CID | 74547 |
| IUPAC Name | (2-chlorophenyl)-(4-fluorophenyl)methanone |
| SMILES | C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)Cl |
Chemical Identifiers
| 1806-23-1 | |
| 234.654 | |
| DODIKYQYCCFWRZ-UHFFFAOYSA-N | |
| (2-chlorophenyl)-(4-fluorophenyl)methanone |
| C13H8ClFO | |
| MFCD00000559 | |
| 74547 | |
| C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)Cl |
Specifications
| 1806-23-1 | |
| C13H8ClFO | |
| 5 g | |
| C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)Cl | |
| 234.654 | |
| 234.65 | |
| Crystalline Powder |
| 62°C | |
| MFCD00000559 | |
| DODIKYQYCCFWRZ-UHFFFAOYSA-N | |
| (2-chlorophenyl)-(4-fluorophenyl)methanone | |
| 74547 | |
| ≥98.0% (GC) | |
| 2-Chloro-4′-fluorobenzophenone |
Safety and Handling
EINECSNumber : (4)-1488
TSCA : Yes