missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4'-fluorobenzophenone 98.0+%, TCI America™

Catalog No. C18605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18605G 5 g
C186025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18605G Supplier TCI America Supplier No. C18605G
Only null left

Chemical Identifiers

CAS 1806-23-1
Molecular Formula C13H8ClFO
Molecular Weight (g/mol) 234.654
MDL Number MFCD00000559
InChI Key DODIKYQYCCFWRZ-UHFFFAOYSA-N
PubChem CID 74547
IUPAC Name (2-chlorophenyl)-(4-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)Cl

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 234.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4′-fluorobenzophenone
EINECSNumber (4)-1488
TSCA Yes