missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzaldehyde 97.0+%, TCI America™

Catalog No. C14651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14651G 1 g
C14655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14651G Supplier TCI America Supplier No. C14651G

Chemical Identifiers

CAS 84194-36-5
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00042527
InChI Key KMQWNQKESAHDKD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-benzaldehyde, benzaldehyde, 2-chloro-4-fluoro, pubchem1433, acmc-209pu2, 2-chloro-4-fluorobezaldehyde, ksc448a5h, 2-chloro 4-fluorobenzaldehyde, 4-fluoro-2-chlorobenzaldehyde, 2-chloro 4-fluoro benzaldehyde, timtec-bb sbb003983
PubChem CID 145024
IUPAC Name 2-chloro-4-fluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(Cl)=C1

Specifications

Melting Point 61°C
Color White
Quantity 1 g
Formula Weight 158.56
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluorobenzaldehyde