missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-4'-fluoroacetophenone 97.0+%, TCI America™

Catalog No. C26435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26435G 5 g
C26431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26435G Supplier TCI America Supplier No. C26435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 700-35-6
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD00042202
InChI Key CSEMGLVHVZRXQF-UHFFFAOYSA-N
Synonym 1-2-chloro-4-fluorophenyl ethanone, 2'-chloro-4'-fluoroacetophenone, 2-chloro-4-fluoroacetophenone, 1-2-chloro-4-fluorophenyl ethan-1-one, ethanone, 1-2-chloro-4-fluorophenyl, 1-acetyl-2-chloro-4-fluorobenzene, pubchem4187, 2-chloro-4-fluoracetophenone, ksc631o2l, 4'-fluoro-2'-chloroacetophenone
PubChem CID 2736507
IUPAC Name 1-(2-chloro-4-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)Cl

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 172.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Chloro-4′-fluoroacetophenone