accessibility menu, dialog, popup

UserName

H3COFCl

CAS RN 700-35-6

IUPAC Name: 1-(2-chloro-4-fluorophenyl)ethan-1-one
Synonyms: 1-(2-chloro-4-fluorophenyl)ethanone
Molecular Weight (g/mol): 172.58
Molecular Formula: C8H6ClFO
InChi Key: CSEMGLVHVZRXQF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results