missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluoro-5-nitrobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C244625G
Specifications
| 2-Chloro-4-fluoro-5-nitrobenzoic Acid | |
| 154°C | |
| C7H3ClFNO4 | |
| 25 g | |
| SYZKAFCPWNFONG-UHFFFAOYSA-N | |
| 219.552 | |
| 219.55 | |
| Crystalline Powder |
| 114776-15-7 | |
| White-Yellow | |
| MFCD04115715 | |
| 2-chloro-4-fluoro-5-nitrobenzoic acid, 2-chloro-4-fluoro-5-nitro-benzoic acid, benzoic acid, 2-chloro-4-fluoro-5-nitro, benzoic acid,2-chloro-4-fluoro-5-nitro, acmc-2099ln, 5-carboxy-4-chloro-2-fluoronitrobenzene, 2,2'-bithiophene-5,8'-diylbis trimethylstannane | |
| C1=C(C(=CC(=C1[N+](=O)[O-])F)Cl)C(=O)O | |
| 7210828 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 114776-15-7 | |
| 219.552 | |
| SYZKAFCPWNFONG-UHFFFAOYSA-N | |
| 7210828 |
| C7H3ClFNO4 | |
| MFCD04115715 | |
| 2-chloro-4-fluoro-5-nitrobenzoic acid, 2-chloro-4-fluoro-5-nitro-benzoic acid, benzoic acid, 2-chloro-4-fluoro-5-nitro, benzoic acid,2-chloro-4-fluoro-5-nitro, acmc-2099ln, 5-carboxy-4-chloro-2-fluoronitrobenzene, 2,2'-bithiophene-5,8'-diylbis trimethylstannane | |
| C1=C(C(=CC(=C1[N+](=O)[O-])F)Cl)C(=O)O |
Safety and Handling
TSCA : No