missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C24465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24465G 5 g
C244625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24465G Supplier TCI America Supplier No. C24465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114776-15-7
Molecular Formula C7H3ClFNO4
Molecular Weight (g/mol) 219.552
MDL Number MFCD04115715
InChI Key SYZKAFCPWNFONG-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-5-nitrobenzoic acid, 2-chloro-4-fluoro-5-nitro-benzoic acid, benzoic acid, 2-chloro-4-fluoro-5-nitro, benzoic acid,2-chloro-4-fluoro-5-nitro, acmc-2099ln, 5-carboxy-4-chloro-2-fluoronitrobenzene, 2,2'-bithiophene-5,8'-diylbis trimethylstannane
PubChem CID 7210828
SMILES C1=C(C(=CC(=C1[N+](=O)[O-])F)Cl)C(=O)O

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 5 g
Formula Weight 219.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrobenzoic Acid