missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-1-iodobenzene, 98%

Catalog No. p-7081843
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1201006 5 g
AAA1201014 25 g
AAA1201022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1201006 Supplier Thermo Scientific Chemicals Supplier No. A1201006
Only null left

CAS: 101335-11-9 | C6H3ClFI | 256.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101335-11-9
Molecular Formula C6H3ClFI
Molecular Weight (g/mol) 256.44
MDL Number MFCD00051774
InChI Key POTCKVPDYXEGSV-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoroiodobenzene, 1-chloro-5-fluoro-2-iodobenzene, benzene, 2-chloro-4-fluoro-1-iodo, 2-chloro-4-fluoro-1-iodo-benzene, pubchem3434, acmc-1bu0g, ksc494m4h, 4-fluoro-2-chloro-iodobenzene, 2-chloro-4-fluoro-iodobenzene, 2-chloro-4-fluoro-1-iodobenzol
PubChem CID 2736559
IUPAC Name 2-chloro-4-fluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(Cl)=C1

Specifications

Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.601
Quantity 5 g
Beilstein 2435080
Sensitivity Light sensitive
Formula Weight 256.45
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluoro-1-iodobenzene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only