missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(dimethylamino)benzaldehyde 98.0+%, TCI America™

Catalog No. c26065g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26065G 5 g
C26061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26065G Supplier TCI America Supplier No. C26065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1424-66-4
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.64
MDL Number MFCD00021051
InChI Key XSQFAWMDRFSIMY-UHFFFAOYSA-N
PubChem CID 74021
IUPAC Name 2-chloro-4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C(Cl)=C1

Specifications

Melting Point 82°C
Color Yellow
Quantity 5 g
Synonym 2-chloro-4-dimethylamino benzaldehyde, benzaldehyde, 2-chloro-4-dimethylamino, benzaldehyde,2-chloro-4-dimethylamino, 2-chloro-4-n,n-bis methyl amino benzaldehyde, 4-dimethylamino-2-chlorobenzaldehyde, acmc-209coh, 3-chloro-n,n-dimethyl-4-formylaniline
Formula Weight 183.64
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-(dimethylamino)benzaldehyde