missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(dimethylamino)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America C26061G
Specifications
| 2-Chloro-4-(dimethylamino)benzaldehyde | |
| 82°C | |
| C9H10ClNO | |
| 1 g | |
| XSQFAWMDRFSIMY-UHFFFAOYSA-N | |
| 2-chloro-4-(dimethylamino)benzaldehyde | |
| 74021 | |
| ≥98.0% (GC,N) |
| 1424-66-4 | |
| Yellow | |
| MFCD00021051 | |
| 2-chloro-4-dimethylamino benzaldehyde, benzaldehyde, 2-chloro-4-dimethylamino, benzaldehyde,2-chloro-4-dimethylamino, 2-chloro-4-n,n-bis methyl amino benzaldehyde, 4-dimethylamino-2-chlorobenzaldehyde, acmc-209coh, 3-chloro-n,n-dimethyl-4-formylaniline | |
| CN(C)C1=CC=C(C=O)C(Cl)=C1 | |
| 183.64 | |
| 183.64 | |
| Crystalline Powder |
Chemical Identifiers
| 1424-66-4 | |
| 183.64 | |
| XSQFAWMDRFSIMY-UHFFFAOYSA-N | |
| 2-chloro-4-(dimethylamino)benzaldehyde |
| C9H10ClNO | |
| MFCD00021051 | |
| 74021 | |
| CN(C)C1=CC=C(C=O)C(Cl)=C1 |
Safety and Handling
TSCA : Yes