missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%

CAS: 3842-55-5 | C15H10ClN3 | 267.716 g/mol

Supplier:  Thermo Scientific Chemicals H3317506

 View more versions of this product

Catalog No. AAH3317506

Only null left
Explore available promotions

Description

Description

A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Chloro-4,6-diphenyl-1,3,5-triazine
138°C to 140°C
MFCD05738885
1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
DDGPPAMADXTGTN-UHFFFAOYSA-N
2-chloro-4,6-diphenyl-1,3,5-triazine
19698
97%
3842-55-5
C15H10ClN3
5 g
Soluble in toluene.
C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3
267.716
267.72

Chemical Identifiers

3842-55-5
267.716
DDGPPAMADXTGTN-UHFFFAOYSA-N
19698
C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3
C15H10ClN3
MFCD05738885
1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
2-chloro-4,6-diphenyl-1,3,5-triazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only