missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%

Catalog No. p-7031912
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3317503 1 g
AAH3317506 5 g
AAH3317514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3317503 Supplier Thermo Scientific Chemicals Supplier No. H3317503
Only null left

CAS: 3842-55-5 | C15H10ClN3 | 267.716 g/mol

A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3842-55-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.716
MDL Number MFCD05738885
InChI Key DDGPPAMADXTGTN-UHFFFAOYSA-N
Synonym 1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
PubChem CID 19698
IUPAC Name 2-chloro-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

Melting Point 138°C to 140°C
Quantity 1 g
Solubility Information Soluble in toluene.
Formula Weight 267.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,6-diphenyl-1,3,5-triazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only