Learn More
2-Chloro-4,6-dinitroaniline, 97%, Thermo Scientific Chemicals
$77.32 - $77.32
Chemical Identifiers
| CAS | 3531-19-9 |
|---|---|
| Molecular Formula | C6H4ClN3O4 |
| Molecular Weight (g/mol) | 217.57 |
| MDL Number | MFCD00007147 |
| InChI Key | LHRIICYSGQGXSX-UHFFFAOYSA-N |
| Synonym | 6-chloro-2,4-dinitroaniline, 2-chloro-4,6-dinitrophenyl amine, benzenamine, 2-chloro-4,6-dinitro, ccris 3112, aniline, 6-chloro-2,4-dinitro, 6-chloro-2,4-dinitrophenylamine, acmc-209ig0, 2,4-dinitro-6-chloroaniline, 3-12-00-01718 beilstein handbook reference, aronis23887 |
| PubChem CID | 19052 |
| IUPAC Name | 2-chloro-4,6-dinitroaniline |
| SMILES | NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2437922
|
Thermo Scientific Chemicals
B2437922 |
100 g |
Each for $77.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3531-19-9 | |
| 217.57 | |
| LHRIICYSGQGXSX-UHFFFAOYSA-N | |
| 19052 | |
| NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O |
| C6H4ClN3O4 | |
| MFCD00007147 | |
| 6-chloro-2,4-dinitroaniline, 2-chloro-4,6-dinitrophenyl amine, benzenamine, 2-chloro-4,6-dinitro, ccris 3112, aniline, 6-chloro-2,4-dinitro, 6-chloro-2,4-dinitrophenylamine, acmc-209ig0, 2,4-dinitro-6-chloroaniline, 3-12-00-01718 beilstein handbook reference, aronis23887 | |
| 2-chloro-4,6-dinitroaniline |
Specifications
| 3531-19-9 | |
| C6H4ClN3O4 | |
| 100 g | |
| 6-chloro-2,4-dinitroaniline, 2-chloro-4,6-dinitrophenyl amine, benzenamine, 2-chloro-4,6-dinitro, ccris 3112, aniline, 6-chloro-2,4-dinitro, 6-chloro-2,4-dinitrophenylamine, acmc-209ig0, 2,4-dinitro-6-chloroaniline, 3-12-00-01718 beilstein handbook reference, aronis23887 | |
| NC1=C(Cl)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O | |
| 217.57 | |
| 217.57 | |
| 2-Chloro-4,6-dinitroaniline |
| 153°C to 155°C | |
| MFCD00007147 | |
| UN2811 | |
| LHRIICYSGQGXSX-UHFFFAOYSA-N | |
| 2-chloro-4,6-dinitroaniline | |
| 19052 | |
| 97% |
Safety and Handling
GHS H Statement
H300-H310-H330-H373-H315-H319
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 222-564-9
RTECSNumber : BX1226000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only