Learn More
2-Chloro-4,6-dimethylpyrimidine, 97%
CAS: 4472-44-0 | C6H7ClN2 | 142.59 g/mol
$205.38 - $205.38
Chemical Identifiers
| CAS | 4472-44-0 |
|---|---|
| Molecular Formula | C6H7ClN2 |
| Molecular Weight (g/mol) | 142.59 |
| InChI Key | RZVPFDOTMFYQHR-UHFFFAOYSA-N |
| Synonym | pyrimidine, 2-chloro-4,6-dimethyl, 2-chloro-4,6-dimethyl-pyrimidine, pubchem6904, acmc-1adw7, 5-23-05-00401 beilstein handbook reference, ksc495i5r, 2-chlor-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethylpyrimidine |
| PubChem CID | 20550 |
| IUPAC Name | 2-chloro-4,6-dimethylpyrimidine |
| SMILES | CC1=CC(=NC(=N1)Cl)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC442550050
|
Thermo Scientific Chemicals
442550050 |
5 g |
Each for $205.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4472-44-0 | |
| 142.59 | |
| pyrimidine, 2-chloro-4,6-dimethyl, 2-chloro-4,6-dimethyl-pyrimidine, pubchem6904, acmc-1adw7, 5-23-05-00401 beilstein handbook reference, ksc495i5r, 2-chlor-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethylpyrimidine | |
| 2-chloro-4,6-dimethylpyrimidine |
| C6H7ClN2 | |
| RZVPFDOTMFYQHR-UHFFFAOYSA-N | |
| 20550 | |
| CC1=CC(=NC(=N1)Cl)C |
Specifications
| 4472-44-0 | |
| 100.0 | |
| White to Yellow | |
| Authentic | |
| C6H7ClN2 | |
| pyrimidine, 2-chloro-4,6-dimethyl, 2-chloro-4,6-dimethyl-pyrimidine, pubchem6904, acmc-1adw7, 5-23-05-00401 beilstein handbook reference, ksc495i5r, 2-chlor-4,6-dimethylpyrimidine, 4,6-dimethyl-2-chloropyrimidine, 2-chloro-4,6-dimethyl-1,3-diazine, 2-chloro-4,6-dimethylpyrimidine | |
| CC1=CC(=NC(=N1)Cl)C | |
| 142.59 | |
| 142.59 | |
| Powder, Crystalline Powder or Low Melting Solid |
| 96.0 | |
| 36°C to 38°C | |
| 210°C to 212°C | |
| 96% min. (GC) | |
| 5 g | |
| RZVPFDOTMFYQHR-UHFFFAOYSA-N | |
| 2-chloro-4,6-dimethylpyrimidine | |
| 20550 | |
| 97% | |
| 2-Chloro-4, 6-dimethylpyrimidine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only