missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile 98.0+%, TCI America™
$72.70 - $361.62
Chemical Identifiers
| CAS | 14237-71-9 |
|---|---|
| Molecular Formula | C8H7ClN2 |
| Molecular Weight (g/mol) | 166.61 |
| MDL Number | MFCD00051676 |
| InChI Key | RETJKTAVEQPNMH-UHFFFAOYSA-N |
| Synonym | 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile |
| PubChem CID | 223261 |
| IUPAC Name | 2-chloro-4,6-dimethylpyridine-3-carbonitrile |
| SMILES | CC1=CC(C)=C(C#N)C(Cl)=N1 |
Chemical Identifiers
| 14237-71-9 | |
| 166.61 | |
| RETJKTAVEQPNMH-UHFFFAOYSA-N | |
| 223261 | |
| CC1=CC(C)=C(C#N)C(Cl)=N1 |
| C8H7ClN2 | |
| MFCD00051676 | |
| 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile | |
| 2-chloro-4,6-dimethylpyridine-3-carbonitrile |
Specifications
| 14237-71-9 | |
| White-Yellow | |
| MFCD00051676 | |
| 3439 | |
| RETJKTAVEQPNMH-UHFFFAOYSA-N | |
| 2-chloro-4,6-dimethylpyridine-3-carbonitrile | |
| 223261 | |
| ≥98.0% (GC) | |
| 2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile |
| 99°C | |
| C8H7ClN2 | |
| 5 g | |
| 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile | |
| CC1=CC(C)=C(C#N)C(Cl)=N1 | |
| 166.61 | |
| 166.61 | |
| Crystalline Powder |
Safety and Handling
TSCA : No