missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile 98.0+%, TCI America™

Supplier:  TCI America C266825G

 View more versions of this product

Catalog No. C266825G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Chloro-4,6-dimethyl-3-pyridinecarbonitrile
99°C
C8H7ClN2
25 g
2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile
CC1=CC(C)=C(C#N)C(Cl)=N1
166.61
166.61
Crystalline Powder
14237-71-9
White-Yellow
MFCD00051676
3439
RETJKTAVEQPNMH-UHFFFAOYSA-N
2-chloro-4,6-dimethylpyridine-3-carbonitrile
223261
≥98.0% (GC)

Chemical Identifiers

14237-71-9
166.61
RETJKTAVEQPNMH-UHFFFAOYSA-N
223261
CC1=CC(C)=C(C#N)C(Cl)=N1
C8H7ClN2
MFCD00051676
2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile
2-chloro-4,6-dimethylpyridine-3-carbonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions