missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. C143310G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C143310G 10 g
C143325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C143310G Supplier TCI America Supplier No. C143310G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13223-25-1
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.584
MDL Number MFCD00274530
InChI Key PBEKEFWBLFBSGQ-UHFFFAOYSA-N
PubChem CID 5152323
IUPAC Name 2-chloro-4,6-dimethoxypyrimidine
SMILES COC1=CC(=NC(=N1)Cl)OC

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 10 g
Formula Weight 174.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,6-dimethoxypyrimidine