accessibility menu, dialog, popup

UserName

H3CONNClOCH3

CAS RN 13223-25-1

IUPAC Name: 2-chloro-4,6-dimethoxypyrimidine
Synonyms: 2-chloro-4,6-dimethoxypyrimidine
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: PBEKEFWBLFBSGQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(Cl)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results